Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55602. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 16.7kD protein; AAG10; aging-associated gene 10 protein; coiled-coil-helix-coiled-coil-helix domain-containing protein 2, mitochondrial; HCV NS2 trans-regulated protein; mitochondria nuclear retrograde regulator 1; mitochondrial nuclear retrograde regulator 1; MIX17 homolog B; NS2TP
Gene Aliases: C7orf17; MIX17B; MNRR1; NS2TP; PARK22
UniProt ID: (Human) Q9Y6H1
Entrez Gene ID: (Human) 51142
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support