Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • FGF-8A Proteins

          Gibco

          Human FGF-8a Recombinant Protein, PeproTech®

          8 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human FGF-8a Recombinant Protein, PeproTech®

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          100-25A-01M

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 7 publications 7 publications

          ELISA standard (ELISA Std)

          -
          View 1 publication 1 publication

          Bioactivity (Bioactivity)

          Assay-dependent
          -
          Product Specifications

          Species

          Human

          Published species

          Human

          Expression System

          CHO cells

          Amino acid sequence

          QHVREQSLVT DQLSRRLIRT YQLYSRTSGK HVQVLANKRI NAMAEDGDPF AKLIVETDTF GSRVRVRGAE TGLYICMNKK GKLIAKSNGK GKDCVFTEIV LENNYTALQN AKYEGWYMAF TRKGRPRKGS KTRQHQREVH FMKRLPRGHH TTEQSLRFEF LNYPPFTRSL RGSQRTWAPE PR

          Molecular weight

          21.2 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 95% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by a cell proliferation assay using balb/c 3T3 cells.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Product Specific Information

          100-00CC-500UG will be provided as 2 x 250 µg (100-00CC-250UG).

          CHO cell-derived Recombinant Human FGF-8a is a 21.2 kDa protein consisting of 182 amino acid residues.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          FGF-8 is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and FGF-8b are identical, unlike human and mouse FGF-8e and FGF-8f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ace; androgen-induced growth factor; cb110; etID309886.13; fb73a06; fgf-8; fgf8; fibroblast growth factor 8 (androgen-induced); heparin-binding growth factor 8; ibd5031

          View more View less

          Gene Aliases: AIGF; FGF-8; HBGF-8; HH6; KAL6

          View more View less

          UniProt ID: (Human) P55075

          View more View less

          Entrez Gene ID: (Human) 2253

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline