Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Proteins & Peptides ›
          • HGF Proteins

          Gibco

          Human HGF Recombinant Protein, PeproTech®

          62 References
          View all (6) HGF proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human HGF Recombinant Protein, PeproTech®

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Testing Data (1)
          Human HGF Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human HGF Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human HGF Protein (100-39H-500UG) in Bioactivity

          Bioassay analysis of Human HGF Recombinant Protein, PeproTech® (Product # 100-39H-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human HGF Protein in Bioactivity (Bioactivity)

          Product Details

          100-39H-500UG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 44 publications 44 publications

          Miscellaneous PubMed (Misc)

          -
          View 10 publications 10 publications

          ELISA standard (ELISA Std)

          -
          View 1 publication 1 publication

          Bioactivity (Bioactivity)

          Assay-dependent
          -

          Cell culture (Cell Clt)

          -
          View 3 publications 3 publications

          Cell differentiation (Cell Diff)

          -
          View 1 publication 1 publication

          Cell stimulation (Cell Stm)

          -
          View 2 publications 2 publications

          Cell expansion (Cell Exp)

          -
          View 1 publication 1 publication
          Product Specifications

          Species

          Human

          Published species

          Chicken, Human, Mouse

          Expression System

          HEK293 cells

          Amino acid sequence

          Alpha chain: KRRNTIHEFK KSAKTTLIKI DPALKIKTKK VNTADQCANR CTRNKGLPFT CKAFVFDKAR KQCLWFPFNS MSSGVKKEFG HEFDLYENKD YIRNCIIGKG RSYKGTVSIT KSGIKCQPWS SMIPHEHSFL PSSYRGKDLQ ENYCRNPRGE EGGPWCFTSN PEVRYEVCDI PQCSEVECMT CNGESYRGLM DHTESGKICQ RWDHQTPHRH KFLPERYPDK GFDDNYCRNP DGQPRPWCYT LDPHTRWEYC AIKTCADNTM NDTDVPLETT ECIQGQGEGY RGTVNTIWNG IPCQRWDSQY PHEHDMTPEN FKCKDLRENY CRNPDGSESP WCFTTDPNIR VGYCSQIPNC DMSHGQDCYR GNGKNYMGNL SQTRSGLTCS MWDKNMEDLH RHIFWEPDAS KLNENYCRNP DDDAHGPWCY TGNPLIPWDY CPISRCEGDT TPTIVNLDHP VISCAKTKQL R Beta chain: VVNGIPTRTN IGWMVSLRYR NKHICGGSLI KESWVLTARQ CFPSRDLKDY EAWLGIHDVH GRGDEKCKQV LNVSQLVYGP EGSDLVLMKL ARPAVLDDFV STIDLPNYGC TIPEKTSCSV YGWGYTGLIN YDGLLRVAHL YIMGNEKCSQ HHRGKVTLNE SEICAGAEKI GSGPCEGDYG GPLVCEQHKM RMVLGVIVPG RGCAIPNRPG IFVRVAYYAK WIHKIILTYK VPQS

          Molecular weight

          79.4 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 95% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by the dose-dependent stimulation of the proliferation of monkey 4MBr-5 cells. The expected ED50 for this effect is 1.0-20.0 ng/ml.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          500 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          Recombinant Human HGF, sourced from HEK293 cells, is a 79.4 kDa polypeptide consisting of 695 amino acid residues. As a result of glycosylation, Recombinant Human HGF migrates with an apparent molecular mass of approximately 68-85 kDa by SDS-PAGE gel, under non-reducing conditions.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatopoeitin A; hepatopoietin-A; HepatopoietinA; HGF; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; scatter factor); unnamed protein product

          View more View less

          Gene Aliases: DFNB39; F-TCF; HGFB; HPTA; SF

          View more View less

          UniProt ID: (Human) P14210

          View more View less

          Entrez Gene ID: (Human) 3082

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Thermo Fisher Scientific India Pvt Ltd.

          Registered office address:
          402, 403, 404, Delphi 'B' Wing,
          Hiranandani Business Park,
          Powai, Mumbai – 400 076

          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100MH2000PTC126872

          Invitrogen BioServices (India) Pvt Ltd

          Registered office address:
          First Technology Place, No 3, EPIP,
          Whitefield, Bangalore – 560 079

          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100KA2004PTC035330

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-crr4t:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0