Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Proteins & Peptides ›
          • Nectin 2 Proteins

          Invitrogen

          Human Nectin-2 (CD112) Fc Chimera Recombinant Protein

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human Nectin-2 (CD112) Fc Chimera Recombinant Protein

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (1)
          Human Nectin-2 (CD112) Fc Chimera Protein
          Group 53 Created with Sketch.
          Human Nectin-2 (CD112) Fc Chimera Protein
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human Nectin-2 (CD112) Fc Chimera Protein (RP-87967)

          SDS-PAGE analysis of Human Nectin-2 (CD112) Fc Chimera (Product # RP-87967). The specification for purity shows: > 95 % (SDS-PAGE). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human Nectin-2 (CD112) Fc Chimera Protein

          Product Details

          RP-87967

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-Dependent
          -

          ELISA standard (ELISA Std)

          Assay-dependent
          -

          Binding assay (Binding)

          Assay-dependent
          -
          Product Specifications

          Species

          Human

          Expression System

          HEK293 cells

          Amino acid sequence

          Human CD112, amino acids Met1-Leu360 (Accession # NP_002847.1) with a C-terminal human IgG1 Fc region

          Tag

          Fc-tag

          Molecular weight

          70-80 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          >95% as determined by SDS-PAGE.

          Endotoxin concentration

          <1.0 EU/µg

          Activity

          1. Measured by its binding ability in a functional ELISA. Immobilized human CD226 at 2 µg/ml (100 µL/well) can bind human CD112 with a linear range of 6.4-800 ng/mL. 2. Immobilized PVRIG Protein, Cynomolgus, Rhesus, Recombinant (His Tag) at 2 µg/ml (100 µL/well) can bind Nectin-2 Protein, Human, Recombinant (hFc Tag), the EC50 of Nectin-2 Protein, Human, Recombinant (hFc Tag)is 0.2-1.2 ng/mL.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.25 mg/mL

          Amount

          1 mg

          Purification

          purified

          Storage buffer

          PBS, pH 7.4, with 5% mannitol, 5% trehalose, 0.01% Tween 80

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          It is recommended that the protein be aliquoted for optimal storage.

          Reconstitute with sterile deionized water to 0.25mg/mL

          Concentration is measured by UV.

          Target Information

          This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; HveB; Nectin cell adhesion molecule 2; Nectin-2; poliovirus receptor-like 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2; PVRL2; unnamed protein product

          View more View less

          Gene Aliases: CD112; HVEB; NECTIN2; PRR2; PVRL2; PVRR2

          View more View less

          UniProt ID: (Human) Q92692

          View more View less

          Entrez Gene ID: (Human) 5819

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline