Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (57%), Rat (57%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62378. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Odorant receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor OR1-55; olfactory receptor TPCR100
Gene Aliases: HSHTPCRX18; HTPCRX18; OR1-55; OST710; TPCR100
UniProt ID: (Human) Q96R27
Entrez Gene ID: (Human) 26245
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support