Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82497. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in nanoseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predominant source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternatively spliced long form; MGC111763; MGC26458; Regulator of G-protein signaling 9; regulator of G-protein signalling 9; RGS9; RGS9L; unnamed protein product
Gene Aliases: PERRS; PERRS1; RGS9; RGS9L
UniProt ID: (Human) O75916
Entrez Gene ID: (Human) 8787
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support