Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • SEMA3A Proteins

          Gibco

          Human Semaphorin 3A Fc Recombinant Protein, PeproTech®

          3 References
          View all (3) SEMA3A proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human Semaphorin 3A Fc Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (1)
          Human Semaphorin 3A Fc Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human Semaphorin 3A Fc Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human Semaphorin 3A Fc Protein (150-17H-100UG) in Bioactivity

          Bioassay analysis of Human Semaphorin 3A Fc Recombinant Protein, PeproTech® (Product # 150-17H-500). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human Semaphorin 3A Fc Protein in Bioactivity (Bioactivity)

          Product Details

          150-17H-100UG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          ELISA standard (ELISA Std)

          Assay-dependent
          View 1 publication 1 publication

          Binding assay (Binding)

          Assay-dependent
          -

          Bioactivity (Bioactivity)

          Assay-Dependent
          -
          Product Specifications

          Species

          Human

          Published species

          Human, Mouse

          Expression System

          CHO cells

          Amino acid sequence

          HHHHHHHHGK NNVPRLKLSY KEMLESNNVI TFNGLANSSS YHTFLLDEER SRLYVGAKDH IFSFDLVNIK DFQKIVWPVS YTRRDECKWA GKDILKECAN FIKVLKAYNQ THLYACGTGA FHPICTYIEI GHHPEDNIFK LENSHFENGR GKSPYDPKLL TASLLIDGEL YSGTAADFMG RDFAIFRTLG HHHPIRTEQH DSRWLNDPKF ISAHLISESD NPEDDKVYFF FRENAIDGEH SGKATHARIG QICKNDFGGH RSLVNKWTTF LKARLICSVP GPNGIDTHFD ELQDVFLMNF KDPKNPVVYG VFTTSSNIFK GSAVCMYSMS DVRRVFLGPY AHRDGPNYQW VPYQGRVPYP RPGTCPSKTF GGFDSTKDLP DDVITFARSH PAMYNPVFPM NNRPIVIKTD VNYQFTQIVV DRVDAEDGQY DVMFIGTDVG TVLKVVSIPK ETWYDLEEVL LEEMTVFREP TAISAMELST KQQQLYIGST AGVAQLPLHR CDIYGKACAE CCLARDPYCA WDGSACSRYF PTAKRATRAQ DIRNGDPLTH CSDLHHDNHH GHSPEERIIY GVENSSTFLE CSPKSQRALV YWQFQRRNEE RKEEIRVDDH IIRTDQGLLL RSLQQKDSGN YLCHAVEHGF IQTLLKVTLE VIDTEHLEEL LHKDDDGDGS KTKEMSNSMT PSQKVWYRDF MQLINHPNLN TMDEFCEQVW KRDRKQRRQR PGHTPGNSNK WKHLQENKKG RNRRTHEFER APRSVGGPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGK

          Tag

          Fc-tag, His-tag

          Molecular weight

          226.2 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 95% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by its ability to bind recombinant rat Neuropilin-1 Fc Chimera in a functional ELISA assay.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          100 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          150-17H-1MG will be provided as 10 x 100 µg (150-17H-100UG).

          150-17H-500UG will be provided as 5 x 100 µg (150-17H-100UG).

          Recombinant Human Semaphorin 3A Fc has a calculated molecular weight of 226.2 kDa and therefore runs above the 200kDa marker by SDS-PAGE analysis under nonreducing conditions. When run under reducing conditions, this protein migrates as three distinct bands that, due to glycosylation, run higher than expected at apparent molecular weights of approximately 120-130 kDa, 90-100 kDa, and 35-40 kDa.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: collapsin 1; MGC133243; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; Sema III; semaphorin D; Semaphorin III; semaphorin L; Semaphorin-3A; unnamed protein product

          View more View less

          Gene Aliases: coll-1; COLL1; HH16; Hsema-I; Hsema-III; SEMA1; SEMA3A; SEMAD; SEMAIII; SEMAL; SemD

          View more View less

          UniProt ID: (Human) Q14563

          View more View less

          Entrez Gene ID: (Human) 10371

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline