Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Proteins & Peptides ›
          • SLIT2 Proteins

          Gibco

          Human Slit2-N Recombinant Protein, PeproTech®

          9 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human Slit2-N Recombinant Protein, PeproTech®

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          150-11-50UG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 5 publications 5 publications

          Miscellaneous PubMed (Misc)

          -
          View 4 publications 4 publications

          Bioactivity (Bioactivity)

          Assay-dependent
          -
          Product Specifications

          Species

          Human

          Published species

          Human, Mouse

          Expression System

          HEK293 cells

          Amino acid sequence

          QACPAQCSCS GSTVDCHGLA LRSVPRNIPR NTERLDLNGN NITRITKTDF AGLRHLRVLQ LMENKISTIE RGAFQDLKEL ERLRLNRNHL QLFPELLFLG TAKLYRLDLS ENQIQAIPRK AFRGAVDIKN LQLDYNQISC IEDGAFRALR DLEVLTLNNN NITRLSVASF NHMPKLRTFR LHSNNLYCDC HLAWLSDWLR QRPRVGLYTQ CMGPSHLRGH NVAEVQKREF VCSGHQSFMA PSCSVLHCPA ACTCSNNIVD CRGKGLTEIP TNLPETITEI RLEQNTIKVI PPGAFSPYKK LRRIDLSNNQ ISELAPDAFQ GLRSLNSLVL YGNKITELPK SLFEGLFSLQ LLLLNANKIN CLRVDAFQDL HNLNLLSLYD NKLQTIAKGT FSPLRAIQTM HLAQNPFICD CHLKWLADYL HTNPIETSGA RCTSPRRLAN KRIGQIKSKK FRCSAKEQYF IPGTEDYRSK LSGDCFADLA CPEKCRCEGT TVDCSNQKLN KIPEHIPQYT AELRLNNNEF TVLEATGIFK KLPQLRKINF SNNKITDIEE GAFEGASGVN EILLTSNRLE NVQHKMFKGL ESLKTLMLRS NRITCVGNDS FIGLSSVRLL SLYDNQITTV APGAFDTLHS LSTLNLLANP FNCNCYLAWL GEWLRKKRIV TGNPRCQKPY FLKEIPIQDV AIQDFTCDDG NDDNSCSPLS RCPTECTCLD TVVRCSNKGL KVLPKGIPRD VTELYLDGNQ FTLVPKELSN YKHLTLIDLS NNRISTLSNQ SFSNMTQLLT LILSYNRLRC IPPRTFDGLK SLRLLSLHGN DISVVPEGAF NDLSALSHLA IGANPLYCDC NMQWLSDWVK SEYKEPGIAR CAGPGEMADK LLLTTPSKKF TCQGPVDVNI LAKCNPCLSN PCKNDGTCNS DPVDFYRCTC PYGFKGQDCD VPIHACISNP CKHGGTCHLK EGEEDGFWCI CADGFEGENC EVNVDDCEDN DCENNSTCVD GINNYTCLCP PEYTGELCEE KLDFCAQDLN PCQHDSKCIL TPKGFKCDCT PGYVGEHCDI DFDDCQDNKC KNGAHCTDAV NGYTCICPEG YSGLFCEFSP PMV

          Molecular weight

          122.35 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 98% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by its ability to inhibit MC3T3/E1 osteoblasts cell differentiation.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          50 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          150-11-1MG will be provided as 5 x 200 µg (150-11-200UG).

          150-11-500UG will be provided as multiple 100 µg (150-11-100UG) and 200 µg (150-11-200UG) vials.

          Recombinant Human Slit2-N is a 1093 amino acid glycoprotein corresponding to the N-terminal portion of the full length Slit2 precursor, and has a calculated, theoretical molecular weight of 122.35 kDa. Due to glycosylation Slit2-N migrates at an apparent molecular weight of approximately 120.0-140.0 kDa by SDS-PAGE analysis under reducing conditions.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: leucine-rich repeat EGF-like repeat secreted protein; secreted ligand for robo receptors; unnamed protein product

          View more View less

          Gene Aliases: SLIL3; Slit-2

          View more View less

          UniProt ID: (Human) O94813

          View more View less

          Entrez Gene ID: (Human) 9353

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline