SimpliAmp™ Thermal Cycler
The SimpliAmp™ Thermal Cycler is an easy-to-use, accurate, and small thermal cycler designed to fit any lab’s needs for everyday PCR. It is designed with reliable Applied Biosystems® engineering, including innovations like the responsive color touch screen and VeriFlex™ technology for simple and accurate optimization.
EVOS® FL Cell Imaging System
The powerful yet easy-to-use EVOS® FL Cell Imaging System provides the flexibility to fit most epifluorescence microscopy applications. Combining precision components with unique design functionality, the EVOS® FL Cell Imaging System has revolutionized fluorescent microscopy.
QuantStudio® 3D Digital PCR System
Now, you can go beyond the limits of real-time PCR with the new QuantStudio® 3D Digital PCR System. A simple and affordable platform, the QuantStudio® 3D Digital PCR System produces absolute quantification data with the precision and sensitivity you need for your more demanding applications. The QuantStudio® 3D Digital PCR System uses a sealed chip technology, providing a streamlined, reliable, and robust method for performing digital PCR. Just load and go.
Countess® II FL Automated Cell Counter
The new Countess® II FL Automated Cell Counter is a benchtop cell assay platform equipped with state-of-the-art optics and image analysis software for rapid assessment of cells in suspension. Now with fluorescence capabilities—bright-field and two optional fluorescence channels—researchers can count cells, monitor fluorescent protein expression, and measure cell viability.
Ion Chef™ Instrument
he Ion Chef™ System* is the next generation of workflow simplification products for the Ion Proton™ and Ion PGM™ systems. The Ion Chef™ System provides automated, high-throughput template preparation and reproducible chip loading for users at any experience level. With typically less than 15 minutes of hands-on time, an intuitive touch-screen interface, and significantly higher productivity than the Ion OneTouch™ 2 System, the Ion Chef™ System makes the Ion Torrent™ sequencing workflow accessible for every lab.
Attune® NxT Acoustic Focusing Cytometer
The Attune® NxT Acoustic Focusing Cytometer is an easy-to-use instrument designed to detect cell size and granularity, as well as fluorescence expression of labeled antibodies, proteins, and functional reagents. The innovative design of the instrument results in precise data and acquisition speeds up to 10x faster than other flow cytometers. The design also eliminates lengthy sample preparation procedures, reduces the time to analyze a file, allows small or limited samples to be analyzed, and minimizes variability in measurements, leading to better resolution of populations. It is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, and microbiological assays.
Qubit® 3.0 Fluorometer
The Qubit® 3.0 Fluorometer is the next generation of the popular benchtop fluorometer that accurately measures DNA, RNA, and protein using the highly sensitive Qubit® quantitation assays. The concentration of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to to the target, which minimizes the effects of contaminants—including degraded DNA or RNA—on the result. The easy-to-use touch screen menus make it easy to select and run the assays you need, with results displayed in just a few seconds
Ion Personal Genome Machine® (PGM™) System
The new Personal Genome Machine (PGM™) System combines semiconductor sequencing technology with natural biochemistry to directly translate chemical information into digital data, democratizing sequencing and making it accessible to virtually any lab or clinic. The system leverages the exponential improvements in the semiconductor industry (known as Moore’s Law) to provide scalability and flexibility for various applications. The system’s use of the simplest, natural sequencing chemistry eliminates the need for expensive optics and reduces complex chemistries to measure natural DNA extension. Direct, real time sequencing detection provides sequencing results typically in less than 3 hours.
Ion Proton™ System
The Ion Proton™ System is the first benchtop sequencing system capable of human-scale genome, exome, or transcriptome sequencing in a few hours—with DNA-to-variants called in a single day.
The system combines semiconductor sequencing technology with natural biochemistry to directly translate chemical information into digital data. By leveraging the exponential improvements in the semiconductor industry (known as Moore’s Law), the Ion Proton™ Sequencing System provides an unprecedented level of scalability and flexibility to support a broad range of high throughput sequencing applications, ranging from human-scale genome to exome to transcriptome sequencing. The system’s use of the simplest natural sequencing chemistry eliminates the need for expensive optics and complex sequencing chemistries, resulting in a highly affordable sequencing system to own and operate. Real-time, direct, electrical detection of sequencing, combined with the enormous amount of computing power in both the Ion Proton™ Sequencer and Ion Proton™ Torrent Server, enables generation of high quality sequencing results from DNA library to variants in a single day.
QuantStudio™ 12K Flex Real Time PCR System
The QuantStudio™ 12K Flex system is a highly flexible, comprehensive real-time PCR platform that leverages the best of Applied Biosystems real-time technology in a single instrument. The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray® plates, TaqMan® array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for your project whether it’s now or in the future, for maximum flexibility and consistency in your research.