Search
SYTOX Green Nucleic Acid Stain is a bright, high-affinity nucleic acid stain that easily penetrates cells with compromised plasma membranes but does not cross the membranes of live cells, making it a useful indicator of dead cells within a population.
First, fix and permeabilize cultured cells with a protocol appropriate for your sample.
|
| SYTOX Green | |
|---|---|
| Excitation/Emission (nm) | 504/523 |
| Standard filter set | GFP |
| EVOS Light Cube | GFP |
| Storage conditions | ≤–20°C |
Multicolor staining of BPAECs. Mitochondria of BPAECs were stained with MitoTracker Red CM-H2XRos. The cells were then fixed, permeabilized, RNase-treated, stained with SYTOX Green Nucleic Acid Stain, and imaged.
For Research Use Only. Not for use in diagnostic procedures.