Search
View additional product information for Pierce™ Colorimetric Protease Assay Kit - FAQs (23263)
4 product FAQs found
Check the scale used for your X-axis. A logarithmic scale should be used for the X-axis (protease enzyme concentration) rather than a linear scale.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
It could be that the sample protease activity is low relative to the range of dilutions used for the standard curve. You can prepare and use additional standard dilutions to ensure that the full range of assay sensitivity is utilized. Alternatively, the sample protease activity is low relative to the protease used to make standard curve. You can prepare a standard curve using the known amounts of the specific protease being tested.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
There are two potential causes for low absorbance values in the unknown samples. The first being that pH and other buffer conditions were not optimal for the protease being tested. You can repeat the assay using buffer conditions that are optimal for the protease being tested and used to make standard curve. Secondly, it is possible that the protease being tested digests the casein substrate more slowly or less fully than the trypsin standard for which the default assay method was developed. You can perform the digestion step for >20 minutes (e.g., 40 mins); if necessary, also perform the TNBSA development step for longer. You can also use the specific substrate for the protease being tested.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
We offer several specialty protein assays including: Protease assay kits (Cat. Nos. 23263 and 23266), Glycoprotein Carbohydrate Estimation Kit (Cat. No. 23260), Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270), Quantitative Peroxide assay kits (Cat. Nos. 23280 and 23285), and Easy-Titer Assay kits for IgG and IgM.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.