Pierce™ Traut's Reagent (2-iminothiolane) - FAQs
View additional product information for Pierce™ Traut's Reagent (2-iminothiolane) - FAQs (26101)
2 product FAQs found
How long and at what temperature can the Pierce Traut's Reagent (2-iminothiolane) (Cat. No. 26101) be stored in acidic solution devoid of primary amines?
We do not recommend storing the Traut’s Reagent in solution. This reagent is more stable in solution than others crosslinkers because hydrolysis takes place at a slower rate. Yet, its hydrolysis will inevitably occur with time if it is in solution. Therefore, to ensure a better crosslinking efficiency, we recommend preparing a fresh solution each time and storing the Traut's reagent as a powder at 4°C. Unfortunately, we do not have stability data of this reagent in a solution.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can you provide the shelf-life for Traut's Reagent (2-iminothiolane)?
Traut's Reagent (2-iminothiolane) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.