Search
View additional product information for AmpFLSTR™ SGM Plus™ PCR Amplification Kit - FAQs (4307133)
1 product FAQs found
It can amplify 10 tetarnucleotide repeat loci including: D3S1358, vWA, D16S539, D2S1338, D8S1179, D21S11, D18S51, D19S433, TH01, FGA and the Amelogenin gender-determining marker in a single PCR amplification.