Search
View additional product information for TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG - FAQs (4366073, 4366072, 4367846, 4364103, 4352042)
2 product FAQs found
It may be technically feasible, but we have not fully validated TaqMan SNP Genotyping assays on the Applied Biosystems 7500 Fast System and therefore cannot recommend it.
You can perform experiments for Absolute Quantitation and Relative Quantitation with fast cycling. However, Allelic Discrimination (SNP) assays and plus/minus assays using internal positive controls cannot be run with fast reagents and protocols.