Search
View additional product information for Pierce™ Dimethylsulfoxide (DMSO), LC-MS Grade - FAQs (85190)
1 product FAQs found
The DMSO with Cat. Nos. 20688 and 85190 have not been tested for cell freezing. However, Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals (Cat. No. J66650-AE, J66650-AK, J66650-AD) is ⋝99.9% pure and can be used for cell freezing.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center