Water For Injection (WFI) for Cell Culture, 200 L - FAQs
View additional product information for Water For Injection (WFI) for Cell Culture - FAQs (A1287306, A1287305)
2 product FAQs found
What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?
These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How is Water for Injection (WFI) for Cell Culture prepared?
Water for Injection (WFI) for Cell Culture is prepared by distillation and filtration (0.1 µm), with no added substances. It is labeled “For Research Use Only of Futher Manufacturing. Not for use in diagnostic procedures”.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.