Water For Injection (WFI) for Cell Culture
Gibco™

Water For Injection (WFI) for Cell Culture

Gibco™ Water For Injection (WFI) for Cell Culture is high-quality water for use as a solvent in the preparation of cell culture media, laboratory reagents, buffers, and bioprocess liquids.
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
200 L
Catalog number A1287306
also known as A12873-06
Price (USD)
1,500.00
Each
Quantity:
200 L
Price (USD)
1,500.00
Each

Gibco™ Water For Injection (WFI) for Cell Culture is high-quality water for use as a solvent in the preparation of cell culture media, laboratory reagents, buffers, and bioprocess liquids.

Some features of WFI for Cell Culture include:

  • Water For Injection and USP/EP quality
  • Gibco quality and performance
  • Can be used in cell culture, molecular biology, and upstream and downstream bioprocessing applications

Water For Injection quality

WFI for Cell Culture meets USP/EP monograph for Water For Injection at point of fill. The water is prepared by distillation and filtration (0.1 μm), with no added substances. Water quality is monitored on a routine basis. Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly.

Gibco quality and performance

The performance and consistency expected from Gibco liquid media products depends on the high-quality water used in manufacturing. This water is packaged in bottles and CX5-14 bioprocess containers as Gibco Water For Injection (WFI) for Cell Culture. In addition to meeting USP/EP standards, each lot is quality tested for sterility, endotoxin, conductivity, pH, osmolality, nitrates, and total organic carbon.

Use in cell culture and molecular biology applications

WFI for Cell Culture can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, diluting buffers, and cleaning and rinsing agents.

Product use

For use as a raw material component in further manufacturing applications.

cGMP manufacturing and quality system

WFI for Cell Culture is manufactured at cGMP-compliant facilities located in Grand Island, New York and Inchinnan, Scotland. Both facilities are certified to ISO 13485 cGMP manufacturing standards.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingUniversal Bag
Purification MethodMembrane-Filtered
Recommended StorageStore at 2°C to 30°C
Shelf Life24 months from date of manufacture
Shipping ConditionRoom Temperature
SterilitySterile
Treatment(s)None
GradeUSP, EP
Quantity200 L
pH4.0 to 7.5
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How is Water for Injection (WFI) for Cell Culture prepared?

Water for Injection (WFI) for Cell Culture is prepared by distillation and filtration (0.1 µm), with no added substances. It is labeled “For Research Use Only of Futher Manufacturing. Not for use in diagnostic procedures”.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.