EZ-Link™ Sulfo NHS-LC-LC-Biotin, No-Weigh™ Format, 10 x 1 mg - FAQs
View additional product information for EZ-Link™ Sulfo NHS-LC-LC-Biotin, No-Weigh™ Format - FAQs (A35358, 21338)
2 product FAQs found
Can you provide the shelf-life for EZ-Link Sulfo-NHS-LC-LC-Biotin?
EZ-Link Sulfo-NHS-LC-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between EZ-Link Sulfo-NHS-LC-Biotin and Sulfo-NHS-LC-LC-Biotin?
EZ-Link Sulfo-NHS-LC-Biotin has a spacer arm length of 22.4 Angstroms while EZ-Link Sulfo-NHS-LC-LC-Biotin has a spacer arm length of 30.5 Angstroms.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.