CTS™ AIM-V™ Medium, without phenol red, without antibiotics, 1 L - FAQs
View additional product information for CTS™ AIM-V™ Medium, without phenol red, without antibiotics - FAQs (A3830801, A3830802, A4672701)
3 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between CTS AIM-V Medium and CTS AIM V SFM?
These two media have comparable performance and same cGMP quality. The main difference is that CTS AIM-V Medium (Cat. Nos. A3830801, A3830802, A4672701) does not contain phenol red and antibiotics. This medium formulation has been used as an ancillary reagent in a therapeutic cancer vaccine approved by FDA.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is CTS AIM-V Medium, without phenol red, without antibiotics ready-to-use?
Yes, CTS AIM-V Medium, without phenol red, without antibiotics is ready-to-use. Additional supplementation with cytokines and growth factors may be required for certain types of cells. Human serum or CTS Immune Cell SR (Cat. Nos. A2596101 and A2596102) can be added to AIM-V medium to promote cell expansion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.