Search
View additional product information for pBlue TOPO™ TA Expression Kit - FAQs (K483101)
1 product FAQs found
There is no intron in the BGH polyA sequence. The BGH polyA signal (bases 1028-1252 in pcDNA3.1 vector) is the sequence that allows for polyadenylation of the RNA transcript. It is described in the following reference: The 3'-flanking sequence of the bovine growth hormone gene contains novel elements required for efficient and accurate polyadenylation. J Biol Chem.1992 Aug 15;267(23):16330-4
The reference states that the 3' untranslated region leading up to the poly adenylation core sequence (AATAAA) contains unique disperse non-consensus elements (consensus elements are poly U and GU-rich tracts) that serve to increase the efficiency of polyadenylation to a high degree, and that a 9-base sequence downstream of the core sequence facilitates efficient cleavage of the RNA strand. This leads to more stable and higher-abundance transcripts.