What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.