Search
Search

QSOX1| Gene Symbol: QSOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332143.1 | NP_001321934.1 | 8 - 9 | 1190 | 130 | |
| NM_101371.5 | NP_172955.1 | 8 - 9 | 1240 | 130 | ||
| NM_179333.2 | NP_849664.1 | 8 - 9 | 1190 | 130 | ||
| GenBank mRNA | AY062528.1 | - | 8 - 9 | 981 | 130 | |
| BT001218.1 | - | 8 - 9 | 866 | 130 | ||
Set Membership: |
|