Search
Search

AT1G16040| Gene Symbol: AT1G16040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332201.1 | NP_001320971.1 | 5 - 6 | 561 | 111 | |
| NM_001332202.1 | NP_001320970.1 | 6 - 7 | 604 | 111 | ||
| NM_001332203.1 | NP_001320969.1 | 4 - 5 | 490 | 111 | ||
| NM_101472.3 | NP_173056.2 | 6 - 7 | 604 | 111 | ||
| GenBank mRNA | AK118838.1 | - | 6 - 7 | 477 | 111 | |
| AK175134.1 | - | 6 - 7 | 477 | 111 | ||
| AK175454.1 | - | 6 - 7 | 477 | 111 | ||
| BT024928.1 | - | 6 - 7 | 465 | 111 | ||
Set Membership: |
|