Search
Search

PAB1| Gene Symbol: PAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035980.1 | NP_001031057.1 | 6 - 7 | 655 | 65 | |
| NM_101512.4 | NP_173096.1 | 7 - 8 | 558 | 65 | ||
| GenBank mRNA | AF043520.1 | - | 7 - 8 | 491 | 65 | |
| AF332467.1 | - | 6 - 7 | 371 | 65 | ||
| AK317085.1 | - | 7 - 8 | 504 | 65 | ||
| AK317410.1 | - | 6 - 7 | 656 | 65 | ||
| AY088628.1 | - | 7 - 8 | 504 | 65 | ||
| Y13176.1 | - | 491 | 65 | |||
Set Membership: |
|