Search
Search

HAC12| Gene Symbol: HAC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198086.1 | NP_001185015.1 | 13 - 14 | 4397 | 91 | |
| NM_001332245.1 | NP_001321740.1 | 13 - 14 | 4637 | 91 | ||
| NM_001332246.1 | NP_001321737.1 | 13 - 14 | 4847 | 91 | ||
| NM_001332247.1 | NP_001321739.1 | 13 - 14 | 4730 | 91 | ||
| NM_001332248.1 | NP_001321738.1 | 14 - 15 | 4574 | 91 | ||
| NM_001332249.1 | NP_001321742.1 | 13 - 14 | 4335 | 91 | ||
| NM_001332250.1 | NP_001321741.1 | 13 - 14 | 4403 | 91 | ||
| NM_001332251.1 | NP_001321736.1 | 13 - 14 | 4683 | 91 | ||
| NM_001332252.1 | NP_001321735.1 | 13 - 14 | 4664 | 91 | ||
| NM_101532.3 | NP_173115.1 | 12 - 13 | 4430 | 91 | ||
Set Membership: |
|