Search
Search

AT1G16800| Gene Symbol: AT1G16800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332261.1 | NP_001319022.1 | 7 - 8 | 1130 | 84 | |
| NM_001332262.1 | NP_001322185.1 | 8 - 9 | 1461 | 84 | ||
| NM_001332263.1 | NP_001322184.1 | 8 - 9 | 1256 | 84 | ||
Set Membership: |
|