Search
Search

ABCG11| Gene Symbol: ABCG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101647.5 | NP_173226.2 | 1 - 2 | 853 | 84 | |
| GenBank mRNA | AY080792.1 | - | 1 - 2 | 632 | 84 | |
| AY150457.1 | - | 1 - 2 | 385 | 84 | ||
Set Membership: |
|