Search
Search

ABCG11| Gene Symbol: ABCG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101647.5 | NP_173226.2 | 6 - 7 | 1765 | 65 | |
| GenBank mRNA | AY080792.1 | - | 6 - 7 | 1544 | 65 | |
| AY150457.1 | - | 6 - 7 | 1297 | 65 | ||
Set Membership: |
|