Search
Search

ARR7| Gene Symbol: ARR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101763.3 | NP_173339.1 | 3 - 4 | 543 | 130 | |
| GenBank mRNA | AB008490.1 | - | 3 - 4 | 340 | 130 | |
| BT024585.1 | - | 3 - 4 | 307 | 130 | ||
Set Membership: |
|