Search
Search

FNR2| Gene Symbol: FNR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084097.1 | NP_001077566.1 | 3 - 4 | 634 | 91 | |
| NM_001332422.1 | NP_001319047.1 | 2 - 3 | 969 | 91 | ||
| NM_101857.6 | NP_173431.1 | 3 - 4 | 726 | 91 | ||
| GenBank mRNA | AK318722.1 | - | 2 - 3 | 856 | 91 | |
| AY062739.1 | - | 3 - 4 | 518 | 91 | ||
| AY114663.1 | - | 3 - 4 | 500 | 91 | ||
Set Membership: |
|