Search
Search

PSBO1| Gene Symbol: PSBO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126055.4 | NP_201458.1 | 1 - 2 | 418 | 113 | |
| GenBank mRNA | AF424626.1 | - | 1 - 2 | 280 | 113 | |
| AY052299.1 | - | 1 - 2 | 280 | 113 | ||
| AY061924.1 | - | 1 - 2 | 246 | 113 | ||
Set Membership: |
|