Search
Search

CHR1| Gene Symbol: CHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126073.3 | NP_201476.1 | 11 - 12 | 1779 | 135 | |
| GenBank mRNA | AY099638.1 | - | 11 - 12 | 1725 | 135 | |
| BT002161.1 | - | 11 - 12 | 1626 | 135 | ||
Set Membership: |
|