Search
Search

sks17| Gene Symbol: sks17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126091.4 | NP_569041.1 | 3 - 4 | 910 | 91 | |
| GenBank mRNA | AY094401.1 | - | 3 - 4 | 761 | 91 | |
| BT000862.1 | - | 3 - 4 | 648 | 91 | ||
Set Membership: |
|