Search
Search

ABA1| Gene Symbol: ABA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126103.2 | NP_201504.2 | 2 - 3 | 819 | 107 | |
| NM_180954.3 | NP_851285.1 | 2 - 3 | 985 | 107 | ||
| GenBank mRNA | AY093145.1 | - | 2 - 3 | 753 | 107 | |
Set Membership: |
|