Search
Search

IRX14-L| Gene Symbol: IRX14-L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126123.5 | NP_201524.1 | 1 - 2 | 785 | 99 | |
| GenBank mRNA | AY070076.1 | - | 1 - 2 | 739 | 99 | |
| KJ138625.1 | - | 1 - 2 | 711 | 99 | ||
Set Membership: |
|