Search
Search

HOS15| Gene Symbol: HOS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126132.4 | NP_201533.1 | 7 - 8 | 1174 | 69 | |
| GenBank mRNA | AY057698.1 | - | 7 - 8 | 1174 | 69 | |
| AY143932.1 | - | 7 - 8 | 1060 | 69 | ||
Set Membership: |
|