Search
Search

HOS15| Gene Symbol: HOS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126132.4 | NP_201533.1 | 9 - 10 | 1460 | 64 | |
| GenBank mRNA | AY057698.1 | - | 9 - 10 | 1460 | 64 | |
| AY143932.1 | - | 9 - 10 | 1346 | 64 | ||
Set Membership: |
|