Search
Search

FRO1| Gene Symbol: FRO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126159.4 | NP_201560.1 | 3 - 4 | 537 | 74 | |
| GenBank mRNA | AK118663.1 | - | 3 - 4 | 428 | 74 | |
| BT003675.1 | - | 3 - 4 | 379 | 74 | ||
Set Membership: |
|