Search
Search

MEE65| Gene Symbol: MEE65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335052.1 | NP_001325195.1 | 10 - 11 | 757 | 127 | |
| NM_001335053.1 | NP_001325196.1 | 10 - 11 | 757 | 127 | ||
| NM_126189.4 | NP_178237.3 | 10 - 11 | 757 | 127 | ||
Set Membership: |
|