Search
Search

MPK17| Gene Symbol: MPK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035862.2 | NP_001030939.1 | 9 - 10 | 1308 | 87 | |
| NM_001035863.1 | NP_001030940.1 | 9 - 10 | 1390 | 87 | ||
| NM_001035864.1 | NP_001030941.1 | 9 - 10 | 1335 | 87 | ||
| NM_126206.4 | NP_178254.2 | 9 - 10 | 1410 | 87 | ||
| GenBank mRNA | AK228266.1 | - | 9 - 10 | 1301 | 87 | |
| AK317182.1 | - | 9 - 10 | 1308 | 87 | ||
| BT006469.1 | - | 8 - 9 | 1224 | 87 | ||
Set Membership: |
|