Search
Search

TOM3| Gene Symbol: TOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126278.3 | NP_027422.1 | 4 - 5 | 713 | 71 | |
| GenBank mRNA | AB036427.1 | - | 4 - 5 | 451 | 71 | |
| AK117222.1 | - | 4 - 5 | 480 | 71 | ||
Set Membership: |
|