Search
Search

ISPD| Gene Symbol: ISPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335124.1 | NP_001325418.1 | 7 - 8 | 776 | 62 | |
| NM_126305.3 | NP_565286.1 | 7 - 8 | 700 | 62 | ||
| GenBank mRNA | AB037876.1 | - | 7 - 8 | 579 | 62 | |
| AF230737.1 | - | 7 - 8 | 573 | 62 | ||
| AK118110.1 | - | 7 - 8 | 604 | 62 | ||
| BT006120.1 | - | 7 - 8 | 573 | 62 | ||
Set Membership: |
|