Search
Search

RNS1| Gene Symbol: RNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126351.3 | NP_178399.1 | 2 - 3 | 477 | 73 | |
| GenBank mRNA | AY072413.1 | - | 2 - 3 | 312 | 73 | |
| AY114710.1 | - | 2 - 3 | 269 | 73 | ||
| DQ446504.1 | - | 2 - 3 | 269 | 73 | ||
| U05206.1 | - | 2 - 3 | 356 | 73 | ||
Set Membership: |
|