Search
Search

AT3G01380| Gene Symbol: AT3G01380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337331.1 | NP_001319441.1 | 9 - 10 | 1833 | 94 | |
| NM_001337332.1 | NP_001326691.1 | 9 - 10 | 1891 | 94 | ||
Set Membership: |
|