Search
Search

CYP38| Gene Symbol: CYP38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125078.1 | NP_001118550.1 | 2 - 3 | 624 | 66 | |
| NM_111014.4 | NP_186797.1 | 2 - 3 | 687 | 66 | ||
| GenBank mRNA | AY039843.1 | - | 2 - 3 | 622 | 66 | |
| AY087781.1 | - | 2 - 3 | 579 | 66 | ||
| AY113168.1 | - | 2 - 3 | 439 | 66 | ||
| AY568524.1 | - | 2 - 3 | 439 | 66 | ||
Set Membership: |
|