Search
Search

KUP3| Gene Symbol: KUP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111071.3 | NP_186854.1 | 7 - 8 | 1217 | 71 | |
| GenBank mRNA | AY062747.1 | - | 7 - 8 | 1214 | 71 | |
| BT003375.1 | - | 7 - 8 | 1015 | 71 | ||
Set Membership: |
|