Search
Search

SKOR| Gene Symbol: SKOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337437.1 | NP_001325630.1 | 3 - 4 | 796 | 176 | |
| NM_001337438.1 | NP_001325629.1 | 3 - 4 | 615 | 176 | ||
| NM_111153.4 | NP_186934.1 | 3 - 4 | 796 | 176 | ||
Set Membership: |
|