Search
Search

CAT4| Gene Symbol: CAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161114.2 | NP_001154586.1 | 11 - 12 | 1771 | 64 | |
| GenBank mRNA | AY062508.1 | - | 11 - 12 | 1684 | 64 | |
| BT008369.1 | - | 11 - 12 | 1469 | 64 | ||
Set Membership: |
|