Search
Search

AAH| Gene Symbol: AAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341398.1 | NP_001329687.1 | 4 - 5 | 660 | 115 | |
| NM_118126.4 | NP_193740.1 | 4 - 5 | 664 | 115 | ||
| GenBank mRNA | BT025334.1 | - | 4 - 5 | 498 | 115 | |
Set Membership: |
|