Search
Search

sks4| Gene Symbol: sks4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118322.4 | NP_193932.1 | 1 - 2 | 378 | 64 | |
| GenBank mRNA | AY081335.1 | - | 1 - 2 | 240 | 64 | |
| BT008899.1 | - | 1 - 2 | 135 | 64 | ||
Set Membership: |
|